• USA Home
  • Z243302 - ACE Squibb separatory funnel with Locktight stopper
EMAIL THIS PAGE TO A FRIEND
Z243302 Aldrich
ACE Squibb separatory funnel with Locktight stopper
capacity 2,000 mL, stopper Joint: ST/NS 45/50
Purchase
Properties
Related Categories Ace, Ace Funnels, Ace Squibb Separatory Funnel with Locktight Stopper, Glass Funnels, Glassware,
Joint   Joint: ST/NS stopper45/50
mfr. no.   Ace Glass 723165
capacity   2,000 mL
Description
General description
European style funnel with new leaktight locking stopper that will not fall out during shaking or transportation. The stopper is secured into a screw-threaded top joint via cap and Viton® O-ring for a positive, leaktight seal. Supplied complete with 1:5 PTFE plug, stopper, cap, and O-ring.
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?