• USA Home
  • Z243388 - Ace sonochemical reaction vessel with horn coupling
EMAIL THIS PAGE TO A FRIEND
Z243388 Aldrich
Ace sonochemical reaction vessel with horn coupling
223 external O-ring (1)
Purchase
Properties
Related Categories Ace, Ace Ultrasonics, Labware, Manufacturer Browser, Sonochemical reaction vessels,
packaging   pkg of 3 ea
mfr. no.   Ace Glass 7855774
Description
General description
Glass reaction vessel with walls tapered inward toward bottom to allow operation with smaller volumes. 250 mL cap. reactor has an externally threaded center neck and three 14/20 side necks. Center neck accommodates the horn coupling to provide a closed system (gas tight reaction with a standard 1/2 in. diam. sonochemical horn) without affecting sonic output. Horn, with external thread, fits into a SS adapter with internal thread and O-ring seal. Adapter is then secured by a #36 Ace-Thred into a PTFE coupling, which, in turn, is threaded onto center neck of reaction vessel. All seals are accomplished with FETFE® O-rings. Two internal O-rings and one external O-ring are required. Order reaction vessel, SS adapter, and PTFE coupling separately.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?