• USA Home
  • Z243418 - Introduction To Peptide Chemistry
EMAIL THIS PAGE TO A FRIEND
Z243418 Aldrich
Introduction To Peptide Chemistry
  •  ISBN-10 0471935328
  •  ISBN-13 978-047193532-2
Purchase
Properties
Related Categories Biochemistry Books, Books and Software, Labware More...
publication info   P.D. Bailey, John Wiley & Sons, Ltd, 1992, 240 pp., soft cover
mfr. no.   Wiley
Description
General description
This concise introduction to peptide chemistry serves as a step-by-step guide, emphasizing the principles of peptide chemistry, with its strong links to physical and organic chemistry and biochemistry. Intended primarily for advanced undergraduates, it reflects the increasing importance of peptide chemistry, particularly in relation to solid-phase synthesis, gene sequencing and x-ray crystallography. Chapters cover amino acids, purification and isolation, amino acid analysis, sequencing and synthesis. A concluding chapter discusses the structure of LH-RH.
Table of Contents
Amino Acids.
Purification and Isolation of Peptides.
Amino Acid Analysis.
Sequencing.
Synthesis.
The Structure of LH-RH.
Appendices.
Index and Abbreviations.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?