• USA Home
  • Z243973 - Modern NMR Spectroscopy: A Guide for Chemists
EMAIL THIS PAGE TO A FRIEND
Z243973 Aldrich
Modern NMR Spectroscopy: A Guide for Chemists
  •  ISBN-10 0198555679
  •  ISBN-13 978-019855567-4
Purchase
Properties
publication info   J. Sanders and B. Hunter, Oxford University Press, 1993, 328 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
This book provides a non-mathematical, descriptive approach to modern NMR spectroscopy, taking examples from organic, inorganic, and biological chemistry. It also contains much practical advice about the acquisition and use of spectra. Starting from the simple ′one pulse′ sequence, the text employs a ′building block′ approach to lead naturally to multiple pulse and two-dimensional NMR. Spectra of readily available compounds illustrate each technique. One- and two-dimensional methods are integrated in three chapters which show how to solve problems by making connections between spins through bonds, through space, or through exchange. There are also chapters on spectrum editing and solids. The final chapter contains a case history which attempts to weave the many strands of the text into a coherent strategy. This second edition reflects the progress made by NMR in the past few years: there is greater emphasis on inorganic nuclei; some two-color spectra are used; the treatment of heteronuclear experiments has moved from direct to ′inverse′ detection; many new examples and spectra have been included; and the literature to early 1992 has been covered. Like the first edition, this work will be highly useful for all NMR spectroscopists: chemists, academic and industrial researchers, and advanced undergraduate and graduate students needing a clear guide to this valuable technology.
Table of Contents
1. Introduction
2. The one-pulse experiment
3. Spin decoupling and difference spectroscopy
4. Multiple pulse experiments
5. The second dimension
6. Connections through bonds
7. Connections through space
8. Connections through chemical exchange
9. Editing
10. Solids
11. Sucrose octa-acetate: a case history
Appendix: Symmetry, non-equivalence, and restricted rotation
Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?