• USA Home
  • Z244422 - Ultrasonic processor
EMAIL THIS PAGE TO A FRIEND
Z244422 Aldrich
Ultrasonic processor
AC/DC input 110 V, 400 W continuous power
Purchase
Properties
Related Categories Branson processors, Labware, Manufacturer Browser, Sonics & Materials, Sonics & Materials Ultrasonic Processors,
mfr. no.   Sonics & Materials VCX400-110V
availability   not available in EU
Description
General description
These 20,000 Hz units are excellent for processing samples from 250 μL to liters.
Features:
• Microprocessor based with digital display of time and power; menu-driven keypad entry of all parameters
• Integral control of sample temperature (1-100 °C ± 1 °C) with automatic shut-off
• Non-volatile memory for up to ten complex protocols
• Timer for up to 10 hours of processing plus on-off pulsing
• Continuously variable amplitude
Unit comes complete with 13 mm threaded horn, calibrated stainless steel temperature probe, tool kit, and instructions.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?