• USA Home
  • Z244899 - Bransonic® ultrasonic cleaner
EMAIL THIS PAGE TO A FRIEND
Z244899 Aldrich
Bransonic® ultrasonic cleaner
Branson 1510MT, AC input 115 V, tank capacity 1.9 L, with mechanical timer
Purchase
Properties
Related Categories Analytical/Chromatography, Branson, Bransonic cleaners, Bransonic ultrasonic cleaner, Laboratory Equipment,
feature   CSA approved
mfr. no.   Branson 1510MT
availability   not available in EU
W × D × H   5 1/2 in. × 4 in. × 6 in.
tank capacity   1.9 L
Description
General description
Heater not included with this model.
These cleaners have impact-resistant plastic enclosures with recessed SS tanks for fewer spills and drips. Beakers or parts should not come into direct contact with bottom of cleaner tank.
Order accessory trays separately.
• Standard models have a single control to activate ultrasonics and set a time limit
• Touch control on digital readout models permits setting and monitoring cleaning cycles up to 99 min, and displays solution temperature in °C
• A degas function is available on these models for immediate use at maximum efficiency
• All units have dependable solid-state circuitry
• Heater temperature 80°C
Legal Information
Bransonic is a registered trademark of Branson Ultrasonics Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?