• USA Home
  • Z244996 - Bransonic® ultrasonic cleaner
EMAIL THIS PAGE TO A FRIEND
Z244996 Aldrich
Bransonic® ultrasonic cleaner
Branson 1510EMTH, AC input 220 V, tank capacity 1.9 L, with mechanical timer and heater
Purchase
Properties
Related Categories Branson, Bransonic cleaners, Bransonic ultrasonic cleaner, Labware, Manufacturer Browser,
feature   CE compliant
mfr. no.   Branson 1510EMTH
W × D × H   5 1/2 in. × 4 in. × 6 in.
tank capacity   1.9 L
Description
General description
These cleaners have impact-resistant plastic enclosures with recessed SS tanks for fewer spills and drips. Beakers or parts should not come into direct contact with bottom of cleaner tank.
Order accessory trays separately.
• Standard models have a single control to activate ultrasonics and set a time limit
• Touch control on digital readout models permits setting and monitoring cleaning cycles up to 99 min, and displays solution temperature in °C
• A degas function is available on these models for immediate use at maximum efficiency
• All units have dependable solid-state circuitry
• Heater temperature 80°C
Legal Information
Bransonic is a registered trademark of Branson Ultrasonics Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?