• USA Home
  • Z245356 - Aldrich® chromatographic column support
EMAIL THIS PAGE TO A FRIEND
Z245356 Aldrich
Aldrich® chromatographic column support
Purchase
Description
General description
12 gauge steel channel frameworks with chemical-resistant Dura-Green coating. Useful for laboratory-scale column support, vacuum filtration, and separation equipment that requires support structures. Includes two channel clamps that will accommodate tri-grip clamps with 1/2 in. diameter rods.
Unassembled; approx. shipping wt. 40 lbs.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?