• USA Home
  • Z245445 - Statistical Methods In Analytical Chemistry
EMAIL THIS PAGE TO A FRIEND
Z245445 Aldrich
Statistical Methods In Analytical Chemistry
  •  ISBN-10 0471584541
  •  ISBN-13 978-047158454-4
Purchase
Properties
publication info   P. Meier, John Wiley & Sons, Ltd, 1993, 344 pp., hard cover
mfr. no.   Wiley
Description
General description
Combines a cookbook approach with the use of PCs and programmable calculators. Contains statistics suitable for the low number of samples, high-pressure situations commonly found in established analytical methods with algorithms to eliminate statistical table handling, sample programs and data sets that allow readers to play with ideas, recognize trends and connections and observe how various equations are linked in decision making. Features applications in GMP/GLP settings. Packed with examples and illustrations.
Table of Contents
Univariate Data.
Bi- and Multivariate Data.
Ancilliary Techniques.
Complex Examples.
Appendices.
References.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?