EMAIL THIS PAGE TO A FRIEND
Z246018 Aldrich
PTFE sleeve
Joint: ST/NS 34/45
Synonym: ptfe sleeves
Purchase
Properties
Related Categories Bel-Art, Bel-Art PTFE Sleeve Stoppers, Bel-Art Stoppers, Joint Sleeves, Labware,
Joint   Joint: ST/NS34/45
packaging   pkg of 6 ea
mfr. no.   Bel-Art
Description
General description
Extremely thin, accurately tapered, slip-on tubes for ground-glass joints prevent seizing and galling. Chemically inert and used without lubricants or adhesives, thus precluding contamination. Temperature range: −100 to 250°C.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?