EMAIL THIS PAGE TO A FRIEND
Z246567 Aldrich

Nickel crucible

low form, capacity 55 mL

Purchase

Properties

Related Categories Analytical Labware, Crucibles, Labware, Valves and Stopcocks
capacity   55 mL
diam. × H   47 mm × 43 mm
thickness   0.035 in.

Description

General description

High resistance to dilute alkalis at low cost. Used for sodium peroxide fusions in the analysis of zirconium. Also used in analyses for niobium, tantalum, or low levels of phosphorus. Order covers separately.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?