• USA Home
  • Z246972 - Magic Touch lab ice pan
EMAIL THIS PAGE TO A FRIEND
Z246972 Aldrich
Magic Touch lab ice pan
midi, red, capacity 4 L
Purchase
Properties
Related Categories Cryoware, Ice and Insulated Containers, Labware, Magic Touch Containers, Manufacturer Browser,
mfr. no.   Bel-Art, M18848-4103
L × W × H   13 in. × 10 in. × 5 in.
capacity   4 L
color   red
Description
General description
Expanded PVC pan is lightweight, leakproof, and unbreakable. Excellent insulative properties make item ideal for containing liquid nitrogen, and ice/saline. Not for use with acetone.
Temp. range: -321 to 200°F. Wall thickness: 3/4 in.
Legal Information
Magic Touch is a trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?