• USA Home
  • Z247006 - Magic Touch ice bucket
EMAIL THIS PAGE TO A FRIEND
Z247006 Aldrich
Magic Touch ice bucket
yellow
Purchase
Properties
Related Categories Cryoware, Ice and Insulated Containers, Labware, Magic Touch Containers, Manufacturer Browser,
mfr. no.   Bel-Art, M18848-4005
parameter   −321-200 °F temperature
capacity   1 gal (4 L)
top diam.   10 1/4 in. (27 cm)
color   yellow
Description
General description
Expanded PVC bucket is light-weight, leak-proof, and unbreakable. Excellent insulative properties make them ideal for containing liquid nitrogen, dry ice/acetone, and ice/saline. One piece with lid.
Legal Information
Magic Touch is a trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z220531 Magic Touch ice bucket, black
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?