EMAIL THIS PAGE TO A FRIEND
Z247030 Aldrich
Coors casserole
capacity 60 mL
Purchase
Properties
Related Categories Coors, Coors Dishes, Dishes; Life-Science, Labware, Life Science Labware & Equipment,
material   circles
  porcelain
sterility   non-sterile
packaging   pack of 6 (2 packs per case)
mfr. no.   Coorstek 60058
capacity   60 mL
diam. × H   70 mm × 35 mm
matrix active group   not treated surface treatment
Description
General description
Porcelain, with lip and flat handle. Glazed inside and out, except for rim and top of handle.
Legal Information
Coors is a trademark of Coorstek, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?