• USA Home
  • Z247391 - Coors table-type fixed plate Buchner funnel
EMAIL THIS PAGE TO A FRIEND
Z247391 Aldrich
Coors table-type fixed plate Buchner funnel
capacity 1,900 mL
Purchase
Properties
Related Categories Buchner, Table-top, Coors, Coors Funnels, Funnels, Labware,
mfr. no.   Coorstek 60282
H   122 mm
capacity   1,900 mL
perforated area diam.   160 mm
plate diam.   186 mm
Description
General description
One-piece porcelain with fixed, perforated plate. Side-outlet for vacuum. Glazed inside and out except for rim.
Legal Information
Coors is a trademark of Coorstek, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
accessory
Product #
Description
Add to Cart
Z756369 Scienceware® Buchner funnel trowel, pkg of 1 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?