• USA Home
  • Z247456 - Coors multi-well plate

EMAIL THIS PAGE TO A FRIEND
Z247456 Aldrich

Coors multi-well plate

suitable for color reaction or spot plates

Purchase

Properties

Related Categories Coors, Coors Microscopy Supplies, Labware, Manufacturer Browser, Microscopy & Staining,
material   porcelain plate
  wells round
  off-white wells
sterility   non-sterile
feature   lid: no
mfr. no.   Coorstek 60427
L × W × thickness   112 mm (4.409 in.) × 92 mm (3.622 in.) × 7 mm (0.275 in.), well dimensions: 21 mm (0.828 in.) diameter x 5 mm (0.220 in.) deep
size   12 wells
color   none
matrix active group   glazed surface treatment

Description

General description

Porcelain. Glazed except for bottom surface. Suitable for color reaction or spot plates. Twelve depressions in total.

Legal Information

Coors is a trademark of Coorstek, Inc.

Price and Availability


Tissue Diagnostics
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?