• USA Home
  • Z247561 - Coors evaporating dishes
EMAIL THIS PAGE TO A FRIEND
Z247561 Aldrich
Coors evaporating dishes
capacity 80 mL
Purchase
Properties
Related Categories Coors, Coors Dishes, Dishes; Life-Science, Labware, Life Science Labware & Equipment,
material   circles
  porcelain
sterility   non-sterile
packaging   pkg of 6 ea
mfr. no.   Coorstek 60198
H   30 mm
capacity   80 mL
top O.D.   80 mm
matrix active group   not treated surface treatment
Description
General description
Porcelain, with lip. Glazed inside and out, except for rim. Max. use temp.: 1,150 °C; limit of use evacuated: 1,300 °C.
Legal Information
Coors is a trademark of Coorstek, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?