• USA Home
  • Z247952 - Stereochemistry Of Organic Compounds
EMAIL THIS PAGE TO A FRIEND
Z247952 Aldrich
Stereochemistry Of Organic Compounds
  •  ISBN-10 0471016705
  •  ISBN-13 978-047101670-0
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   E.L. Eliel, John Wiley & Sons, Ltd, 1994, 1267 pp., hard cover
mfr. no.   Wiley
Description
General description
Long awaited by the chemistry community, here is the most important work on stereochemistry--the study of molecules in 3D--by the leading authorities in the field. It is the first comprehensive review of this important field in 30 years, since Eliel′s original groundbreaking work, Stereochemistry of Carbon Compounds, was published. The book provides a thorough, organized treatment of modern stereochemistry as applied to organic compounds, covering both basic principles and practical aspects. Includes over 1,000 illustrations and chemical structures, numerous tables including ones on conformational energies and physical properties, over 2,500 references, and a glosssary of terms.
Table of Contents
Structure.
Stereoisomers.
Symmetry.
Configuration.
Properties of Stereoisomers: Stereoisomer Discrimination.
Separation of Stereoisomers: Resolution, Racemization.
Heterotopic Ligands and Faces (Prostereoisomerism, Prochirality).
Stereochemistry of Alkenes.
Conformation of Acyclic Molecules.
Configuration and Conformation of Cyclic Molecules.
Stereoselective Synthesis.
Chiroptical Properties.
Chirality in Molecules Devoid of Chiral Centers.
Glossary.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?