• USA Home
  • Z248231 - J-KEM® high power temperature controller
EMAIL THIS PAGE TO A FRIEND
Z248231 Aldrich
J-KEM® high power temperature controller
Model, 250, -200-250 °C temperature
Purchase
Properties
Related Categories Heating, Heating Controllers, J-KEM, Labware, Manufacturer Browser,
AC input   120 V / 1800 watts, 15 A (model 260/Timer)
feature   thermocouple type T
mfr. no.   Model, 250
measuring range   temperature, accuracy: 0.1 °C, (1 mL to 22 L)
  -200-250 °C temperature
output   3
Description
General description
Research grade temperature controller features auto-tune, PID process controllers with solid-state output circuitry and J-KEM′s Power Control computer that virtually eliminates temperature overshoot of the set temperature. Regulates any piece of equipment or volume to 0.1 °C.
• Regulates heating mantles, hot plates, ovens, heating blocks, etc.
• Safe 100% solid-state electronics, double fused, unbreakable thermocouple probe, automatic over-temperature circuit
• Programmable ramp-to-set point temperature control on the 200-Series model
• Free windows software for PC control and data logging to excel or text file. Implements multi-step temperature ramping and real-time strip chart display
• Both USB 2.0 and RS232 communication ports for PC control and data logging
• Over- / Under- temperature alarm
• Powers mantles upto 22 L
Models include: a 1/8 in.. diam. × 12 in. L, PTFE-coated, SS thermocouple probe with a 10 ft extension cable and PTFE probe adapter with 14/20 joint.
without timer
Legal Information
J-KEM is a registered trademark of J-KEM Electronics, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?