• USA Home
  • Z248452 - Cyclization Reactions
EMAIL THIS PAGE TO A FRIEND
Z248452 Aldrich
Cyclization Reactions
  •  ISBN-10 0849378613
  •  ISBN-13 978-084937861-4
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   C. Thebtaranonth and Y. Thebtaranonth, CRC Press, 1993, 384 pp., hard cover
mfr. no.   CRC Press
Description
General description
• Covers the basic principles of cyclization chemistry, emphasizing practical applications
• Summarizes various methodologies used in cyclization reactions
• Extensive use of equations, schemes, tables, and figures enables quick understanding of the material
• Convenient selection of reactions and conditions in synthesis
• More than 2,600 structures illustrate key concepts
Cyclization Reactions provides a quick update of the latest advances in cyclization reactions. It covers the basic principles of cyclization chemistry, emphasizing practical applications. Chapters are organized according to the different cyclization intermediates-cationic, radical, anionic, and metal complex intermediates. The last chapter covers macrolactonization, vicinal tricarbonal, and Bergman (enediyne) reactions, which are of particular interest today.
More than 2,600 structures illustrate key concepts throughout the book. Various cyclizations are organized into mechanistic groups to help researchers choose and change between methods when searching for maximum efficiency in synthesis. Critical coverage of the literature up to 1992 is provided.
Cyclization Reactions is essential reading for anyone involved in the synthesis of ring compounds or who is seeking a rapid overview of the field. Newcomers as well as experienced researchers will benefit from this book. It also is excellent reference material for students at the advanced undergraduate and graduate levels.
Other Notes
Volume 1 of the Series: New Directions in Organic & Biological Chemistry
Table of Contents
Cyclization Reactions.
Cationic Cyclization.
Radical Cyclization.
Anionic Cyclization.
Metal-Catalyzed Cyclization.
Miscellaneous Cyclizations.
References.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?