• USA Home
  • Z248495 - Polymer Syntheses, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z248495 Aldrich
Polymer Syntheses, 2nd ed.
Volume 2
  •  ISBN-10 0126185123
  •  ISBN-13 978-012618512-6
Purchase
Properties
Related Categories Books and Software, Labware, Polymer Chemistry Books More...
publication info   S. Sandler and W. Karo, Academic Press, 1993, 448 pp., case bound
mfr. no.   Elsevier
Description
General description
Provides detailed laboratory instructions for the synthesis of polymers.
Table of Contents
Urea, Melamine, Benzoguanamine-Aldehyde Resins (Amino Resins or Aminoplasts).
Phenol-Aldehyde Condensations.
Epoxy Resins.
Silicone Resins (Polyorganosiloxanes or Silicones).
Alkyd Resins.
Polyacetals and Poly(vinyl acetals).
Poly(vinyl ethers).
Poly(N-Vinylpyrrolidone).
Polymerization of Acrylic Acids and Related Compounds.
Poly(vinyl chloride).
Miscellaneous.
Subject Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?