• USA Home
  • Z249130 - Radiation Curing of Polymeric Materials
EMAIL THIS PAGE TO A FRIEND
Z249130 Aldrich
Radiation Curing of Polymeric Materials
  •  ISBN-10 0841217300
  •  ISBN-13 978-084121730-0
Purchase
Properties
publication info   C. Hoyle and J. Kinstle, ed., American Chemical Society, 1990, 567 pp.
mfr. no.   American Chemical Society
Description
General description
A review of recent developments in photoinitiators, monomers, photodegradation, cationic polymerization, and laser and electron beam initiation.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?