• USA Home
  • Z249327 - Stronghold lattice system
EMAIL THIS PAGE TO A FRIEND
Z249327 Aldrich
Stronghold lattice system
frame size 60 cm × 60 cm
Purchase
Properties
Related Categories Chemistry Equipment and Labware, Clamps and Supports, Harvard Apparatus, Labware, Manufacturer Browser,
description   7 rods of 60 cm
feature   no. of bases 4
  no. of clamps 8
  no. of hinged adapters 2
mfr. no.   Harvard Apparatus
frame size   60 cm × 60 cm
Description
General description
Stronghold clamps and connectors are made of corrosion-resistant, injection molded nylon with fiberglass reinforcement for lightweight and strength. The brilliant red and white color of the hardware is an integral part of the product and will not fail in use like painted or plated coatings. Basic Stronghold kits can be expanded by ordering accessory clamps, adapters, and support bases.
Kit components:
• Lattice rods- 1/2 in. (13 mm) diam. hollow SAE T304 SS tubes with 1.6 mm wall, plugged with red nylon end caps
• Lattice clamps- two open sides set at right angles that lock 1/2 in. (13 mm) diam. lattice rods in place via 5 mm thread 1/2 in. L Allen set screws
• Lattice bases- 2 1/2in. (64 mm) diam. base has three recessed screw holes for mounting on bench or wall; female socket with locking thumbscrew accepts rods up to 1/2 in. (13 mm) diam.
• Male-female hinged adapters- permit rotation of support legs for benchtop/wall installations
All kits include bench/bench-wall mounting assemblies for installing lattice to benchtop alone or with the rear legs swiveled up and attached to a wall at any point up to 90°. Allen screw wrench also included.
Legal Information
Stronghold is a trademark of Harvard Apparatus
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?