• USA Home
  • Z249394 - Aspirator pump, Büchi® Model B-169
EMAIL THIS PAGE TO A FRIEND
Z249394 Aldrich
Aspirator pump, Büchi® Model B-169
AC input 220 V AC50 Hz
Purchase
Properties
mfr. no.   Brinkman 15-50-802-7
W × D × H   14 in. × 9 in. × 15 in.
Description
General description
Safely aspirates corrosive and volatile solvents without cold or chemical traps. Vapors are condensed and diluted in an internal 10 L PP reservoir for easy disposal. Two vacuum ports equipped with quick coupling connectors and check valves, each with a free air displacement of 12 to 15 L/min.
Achieves a vacuum of 20 mbar at a water temperature of 20 °C. Circulating pump is quiet and requires no oil or filter/trap replacements. Use with rotary evaporators, distillation or filtration apparatus, and electrophoresis gel driers. CE compliant.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?