• USA Home
  • Z249548 - Micro diamond knife
EMAIL THIS PAGE TO A FRIEND
Z249548 Aldrich
Micro diamond knife
Purchase
Properties
Related Categories Knives and Cutters, Labware More...
Description
General description
For the preparation of laboratory samples including those for infrared microanalysis. Has a 60° angled edge with a reinforced beveled tip that is 50 times sharper than a razor blade. Lightweight pen design for safe and convenient use.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z676225 Micro-cutting tool set, for microscopy scale applications
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?