EMAIL THIS PAGE TO A FRIEND
Z249556 Aldrich
Mini-plane
pocket size, for preparing microscopy samples
Purchase
Properties
Related Categories Knives and Cutters, Labware More...
Description
General description
Has a hardened tool-steel blade for scraping off thin sections of polymers, laminates, coatings, and tissues. The thickness of the slice is adjustable with an Allen key. Pocket size with a sturdy wooden handle. Excellent for the preparation of samples for microscopy.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?