• USA Home
  • Z249580 - Whatman® VACU-GUARD® inline filter devices

EMAIL THIS PAGE TO A FRIEND
Z249580 Aldrich

Whatman® VACU-GUARD® inline filter devices

VACU-GUARD 150, molecular sieve, 1/pk

Synonym: filter media, filtration system, inline filtration device

Purchase

Properties

Related Categories Labware, Manufacturer Browser, Pumps, Vacuum Pump Accessories, Vacuum Pumps,
material   PEEK/PTFE filter membrane (PFTE filteration media)
  polypropylene housing
feature   flow rate 250 L/min (1.45 psi differential pressure)
  flow rate 570 L/min (7.25 psi differential pressure)
  Trap media (Molecular sieves, 4Å)
packaging   pack of 1
mfr. no.   Whatman Article No., 28420218 (US reference)
  Whatman, 6722-1003
size   9.3 in. (236 mm) , overall
weight   363 g

Description

General description

Whatman® VACU-GUARD® Inline Filter Devices

The easy-to-use VACU-GUARD inline filter devices help to confine and isolate contaminants in vacuum systems and protect your laboratory.

Features and benefits:
• Excellent for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols
• Designed for in-line use with stepped barb connections for 10 to 12 mm ID hose
• Available with choice of chemical trap: activated carbon, molecular sieve or desiccant
• Membrane retains 99.99% of airbone particles greaterthan 0.1 μm
• Features hydrophobic PTFE membrane

Applications:
• Vacuum pump protection
• Activated carbon removes organic vapors and radioactive particles
• Molecular sieve for use with organic and alkaline air streams
• Desiccant for use with high velocity acidic air streams
• Removes a potential health hazard from the work place

VACU-GUARD 150 Activated carbon
• Chemical Trap Media: Activated carbon
• Filter media: PTFE
• Surface area or weight (nominal): 82,000m2 (carbon)
• Connectors - inlet/outlet: Hose barb/stepped barb

VACU-GUARD 150 Desiccant
• Chemical Trap Media: Anhydrous calcium sulfate
• Filter media: PTFE
• Surface area or weight (nominal): 318 g (desiccant)
• Connectors - inlet/outlet: Hose barb/stepped barb

VACU-GUARD 150 Molecular sieve
• Chemical Trap Media: Silico aluminate zeolite
• Filter media: PTFE
• Surface area or weight (nominal): 318 g (zeolite)
• Connectors - inlet/outlet: Hose barb/stepped barb

Other Notes

Global SIAL customers (excluding Japan) can now order any product from the GE Healthcare Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.

The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.

This product has been replaced by WHA67221003-ALDRICH | Whatman VACU-GUARD inline filter devices, VACU-GUARD 150, molecular sieve, 1/pk

Legal Information

VACU-GUARD is a registered trademark of Arbor Technologies, Inc.

Whatman is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

WHA67221003 Whatman® VACU-GUARD® inline filter devices, VACU-GUARD 150, molecular sieve, 1/pk

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?