• USA Home
  • Z250368 - Asymmetric Catalysis In Organic Synthesis
EMAIL THIS PAGE TO A FRIEND
Z250368 Aldrich
Asymmetric Catalysis In Organic Synthesis
  •  ISBN-10 0471572675
  •  ISBN-13 978-047157267-1
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   R. Noyori, John Wiley & Sons, Ltd, 1994, 400 pp., hard cover
mfr. no.   Wiley
Description
General description
Deals with basic principles of asymmetric catalysis focusing on its synthetic significance. Covers homogeneous asymmetric hydrogenation, asymmetric catalysis via chiral metal complexes, heterogeneous catalysis and non-organometallic catalysis. The collection of a range of stereoselective reactions illustrate various strategies and methodologies as well as their general utility.
Table of Contents
Homogeneous Asymmetric Hydrogenation.
Enantioselective Isomerization of Olefins.
Asymmetric Catalysis via Chiral Metal Complexes: Selected Examples.
Enantioselective Addition of Organometallic Reagents to Carbonyl Compounds: Chirality Transfer, Multiplication, and Amplification.
Three-Component Synthesis of Prostaglandins.
Asymmetric Catalysis with Purely Organic Compounds.
Heterogeneous Asymmetric Catalysis.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?