• USA Home
  • Z250856 - Oxford Chemistry Primers
EMAIL THIS PAGE TO A FRIEND
Z250856 Aldrich
Oxford Chemistry Primers
No. 12: Organometallics 1 Complexes with Transition Metal-Carbon Sigma Bonds
  •  ISBN-10 0198557507
  •  ISBN-13 978-019855750-0
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   M. Bochmann, Oxford University Press, 1994, 96 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
The field of organometallic chemistry has seen explosive growth over the last forty years. On a fundamental level, new structural and bonding concepts have been discovered, while applications range from catalysis to new synthetic methods. This succinct text outlines the main classes of transition metal organometallic complexes and introduces the reader to the chemistry of compounds with metal-carbon bonds: metal carbonyls, metal alkyls, and metal alkylidenes and alkylidynes. The synthetic methods leading to each class of compounds are illustrated with pertinent examples, followed by the discussion of characteristic structures and reactivity patterns. The book stresses general principles and relates the material to specific applications such as catalytic processes. This book is ideal for supplying a quick overview of the discipline to students of chemistry
Table of Contents
1. A Few Basics
2. Metal Carbonyl Complexes
3. Metal Alkyl Complexes
4. Alkylidene and Alkylidyne Complexes
Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?