• USA Home
  • Z250872 - InsulEze® insulating kit
EMAIL THIS PAGE TO A FRIEND
Z250872 Aldrich
InsulEze® insulating kit
Kit A
Purchase
Properties
Related Categories Glas-Col, Heating, Heating Tapes, InsulEze insulating kit, Labware,
mfr. no.   Glas-Col 100GIK210
Description
General description
400 °C, 1/32 in. thick fiberglass tape in the following widths and lengths: 1 in. × 10 ft, 3 in. × 10 ft, and 6 in. × 5 ft. Also includes 60 in. of 1 in. wide Velcro loop tape and 40 in. of 1 in. wide Velcro hoop tape.
Make insulating less time-consuming, awkward, and unsightly in the lab. Use for heat or cold shielding, insulating over heaters, or as shock absorber. Insulating materials can be cut and shaped to almost any size with minimum fraying of the edges. Material is non-hazardous, non-flammable, and inert to most chemicals.
Legal Information
InsulEze is a registered trademark of Glas-Col Laboratory Products
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?