| Related Categories | Glas-Col, Heating, Heating Tapes, InsulEze insulating kit, Labware, |
| mfr. no. | Glas-Col 100GIK220 |
250 °C, 15 W × 30 L ×
Make insulating less time-consuming, awkward, and unsightly in the lab. Use for heat or cold shielding, insulating over heaters, or as shock absorber. Insulating materials can be cut and shaped to almost any size with minimum fraying of the edges. Material is non-hazardous, non-flammable, and inert to most chemicals.
InsulEze is a registered trademark of Glas-Col Laboratory Products