• USA Home
  • Z251674 - Destruction of Hazardous Chemicals in the Laboratory, 2nd Ed.
EMAIL THIS PAGE TO A FRIEND
Z251674 Aldrich
Destruction of Hazardous Chemicals in the Laboratory, 2nd Ed.
  •  ISBN-10 047157399X
  •  ISBN-13 978-047157399-9
Purchase
Properties
Related Categories Books and Software, Labware, Safety Books More...
publication info   G. Lunn and E.B. Sansone, John Wiley & Sons, Ltd, 1994
mfr. no.   Wiley
Description
General description
An updated collection of detailed procedures used to degrade and dispose of a broad range of hazardous chemicals whether in bulk quantities, solutions in various solvents or spills. This edition features new chapters on the removal of metal ions and biological stains from solution; the degradation of mycotoxins, enzyme inhibitors, polycyclic heterocyclic hydrocarbons and highly reactive reagents such as butyllithium, peracids and phosgene as well as the use of much less hazardous reagents including molecular sieves. A list of hazardous compounds indexed by name, molecular formula and CAS Registry Number offers ready access to the data.
Table of Contents
Partial table of contents:
Acid Halides and Anhydrides.
Aflatoxins.
Alkali and Alkaline Earth Metals.
Antineoplastic Alkylating Agents.
Aromatic Amines.
Azides.
Biological Stains.
Boron Trifluoride and Inorganic Fluorides.
Butyllithium.
Calcium Carbide.
Carbamic Acid Esters.
Cisplatin.
Citrinin.
Complex Metal Hydrides.
Cycloserine.
Dimethyl Sulfate and Related Compounds.
Haloethers.
Heavy Metals.
Mercury.
Mitomycin C. Nitrosourea Drugs.
Ochratoxin A. Organic Nitriles.
Patulin.
Peroxides and Hydroperoxides.
Picric Acid.
Protease Inhibitors.
Sodium Amide.
Sulfur-Containing Compounds.
Uranyl Compounds.
Appendices.
Indexes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?