• USA Home
  • Z251844 - Michel-Miller column filter disc
EMAIL THIS PAGE TO A FRIEND
Z251844 Aldrich
Michel-Miller column filter disc
Ace-Thred #15
Purchase
Description
General description
Use at either end of column. Filter disc sizes 7 and 11 are made from specially pure, very uniform, highly-absorbent paper. Order adapter separately.
Price and Availability
All labs need water
Non-Sterile Millex Syringe Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?