• USA Home
  • Z251852 - Michel-Miller column coupling
EMAIL THIS PAGE TO A FRIEND
Z251852 Aldrich
Michel-Miller column coupling
Ace-Thred #15-15
Purchase
Description
General description
PTFE with 316 SS Liner connects M-M columns when packing. Makes a leaktight seal without O-rings. Male Ace Thred matches thread size of columns.
Price and Availability
All labs need water
Non-Sterile Millex Syringe Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?