• USA Home
  • Z251941 - Catalysis Of Organic Reactions by Supported Inorganic Reagents
EMAIL THIS PAGE TO A FRIEND
Z251941 Aldrich
Catalysis Of Organic Reactions by Supported Inorganic Reagents
  •  ISBN-10 0471185604
  •  ISBN-13 978-047118560-4
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   J. Clark, John Wiley & Sons, 1994, 126 pp., hard cover
mfr. no.   Wiley
Description
General description
The book will provide an up-to-date guide and review on a new form of heterogeneous catalysis which will prove to be increasingly important to the chemical industry in the light of new and future environmental legislation. The book will highlight chemical processes where present technology is likely to prove increasingly unacceptable due to associated problems of hazardous waste and/or low process efficiency. It′s purpose is to reveal these problems and possible solutions and to introduce the readers to this new form of catalysis. It will prove to be especially important to fine/speciality/intermediate chemicals manufacturing companies who have little experience of heterogenous catalysis and to researchers interested in chemical aspects of ′Clean Technology′. The book will help to disclose the opportunities for researchers and companies in developing new ′environmentally friendly′ chemical processes
Table of Contents
Supported Reagents.
Characterization of Supported Reagents.
Reactions Catalyzed by Solid Acids.
Reactions Catalyzed by Solid Bases.
Oxidations Catalyzed by Supported Reagents.
Other Reactions Catalyzed by Supported Reagents.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?