• USA Home
  • Z251984 - CHEM-DRY® integrated chemical dryer
EMAIL THIS PAGE TO A FRIEND
Z251984 Aldrich
CHEM-DRY® integrated chemical dryer
AC input 100 - 120 V
Purchase
Description
General description
Dries samples using heat, vacuum, and desiccant. Precise digital control simultaneously displays both "set" and "actual" temperatures from ambient to 175 °C with 1 °C resolution. Temperature ripple is only 1 °C at all temperature settings. A built-in thermocouple automatically maintains selected temperature. Oven features a borosilicate glass sample-viewing port and an integrated safety guard to prevent touching hot parts. Modular design permits the removal and storage of dried samples in sealed drying tubes. Process samples in rapid succession or service multiple users. Order drying tube assembly separately.
Legal Information
CHEM-DRY is a registered trademark of Barnstead International
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z563080 Büchi® glass oven for drying, B-585, AC input 115 V
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?