• USA Home
  • Z252085 - Oxford Chemistry Primers
EMAIL THIS PAGE TO A FRIEND
Z252085 Aldrich
Oxford Chemistry Primers
No. 18: Oscillation, Waves and Chaos in Chemical Kinetics
  •  ISBN-10 0198558449
  •  ISBN-13 978-019855844-6
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   S. Scott, Oxford University Press, 1994, 96 pp.
mfr. no.   Oxford University Press
Description
General description
The phenomena of oscillations, travelling waves, and chaos in reacting chemical systems began as curiosities but now support an active, international research field. This book shows how these "exotic" patterns arise from the underlying chemical mechanisms. The origin of"chemical feedback" is revealed using three example reactions: the isolate-reductant (Landolt) reaction, the Belousov-Zhabotinsky reaction, and the combustion of hydrogen. Thermal feedback is also discussed. These mechanisms lead to clock reactions and travelling reaction fronts, thermal runaway, and flames; to oscillations and excitability; to target patterns, spiral and scroll waves; to bistability, ignition, extinction, and hysteresis; and to complex oscillations and chaos in flow reactors. These phenomena are related to important processes in biology, including the development of cardiac arrhythmias, nerve signal transmission, and animal coat patterning
Table of Contents
1. Introduction to basic concepts
2. Clocks and fronts
3. Resetting the clock: oscillations
4. Targets, spirals and scrolls
5. Bistability: steady states in flow reactors
6. Oscillatory reactions in flow systems
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?