• USA Home
  • Z252093 - Oxford Chemistry Primers
EMAIL THIS PAGE TO A FRIEND
Z252093 Aldrich
Oxford Chemistry Primers
No. 19: Atomic Spectra
  •  ISBN-10 0198556888
  •  ISBN-13 978-019855688-6
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   T. Softley, Oxford University Press, 1994, 96 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
This primer provides a systematic and rigorous introduction to the spectra and electronic structure of atoms in the gas phase. Throughout the book, the author explains observed spectra in terms of underlying quantum mechanical principles while at the same time illustrating experimental aspects and chemical applications. The book begins with a review of elementary quantum mechanics as applied to spherically symmetric problems and a discussion of electromagnetic radiation properties. It goes on to detail the spectra of hydrogenic atoms, the alkali metals, the helium atoms, and many-electron atoms at all stages. Topics such as spin-orbit coupling, the Zeeman effect, spin-correlation and the Pauli Principle, Russel-Saunders, and jj-coupling are included. Modern applications and techniques such as lasers and cooling of atoms are briefly discussed. Many topics are covered at two levels in separate sections, so that a newcomer can gain qualitative understanding by omitting some sections and then return later for a more detailed quantitative picture
Table of Contents
1. Quantum mechanics and light
2. The structure and spectrum of the hydrogen atom
3. The spectra of the alkali metals
4. The spectrum of the helium atom
5. The spectra of many-electron atoms
Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?