• USA Home
  • Z252107 - Oxford Chemistry Primers
EMAIL THIS PAGE TO A FRIEND
Z252107 Aldrich
Oxford Chemistry Primers
No. 20: Chemical Aspects of Biosynthesis
  •  ISBN-10 0198556764
  •  ISBN-13 978-019855676-3
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   J. Mann, Oxford University Press, 1995, 96 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
This introductory text describes and explains the chemistry involved in the biosynthesis of secondary metabolites. The book emphasizes the pharmacological and toxicological significance of the compounds, and the final chapter reviews their ecological role. Suggestions for further reading are provided
Table of Contents
1. Introduction
2. Fatty acids and derivatives
3. Polyketides
4. Terpenoids and steroids: the isoprenoids
5. The shikimic acid pathway: biosynthesis of phenols, lignans, flavonoids, etc.
6. Alkaloids
7. Ecological chemistry
References and further reading
Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?