EMAIL THIS PAGE TO A FRIEND
Z252425 Aldrich
Wheaton vial rack
22 mm tubes and 5 or 10 mL headspace autosampler vials, no. of holes, 36 (3 × 12)
Purchase
Properties
Related Categories Alcan Packaging, Alcan Packaging Vials & Caps, Analytical/Chromatography, Labware, Manufacturer Browser,
feature   no. of holes 36 (3 × 12)
packaging   pkg of 5 ea
L × W × H   9.1 cm × 32.2 cm × 2.8 cm
Description
General description
Chemically resistant polypropylene racks have corner posts to ensure stability when stacking empty. Autoclavable at 121°C for 20min. Vial grid is alphanumerically indexed for easy sample identification.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
23201 Space Saver storage racks, for use with 21-23 mm diameter vial (36 vials per rack), well opening 23.1 mm, pkg of 5 ea
related product
Product #
Description
Add to Cart
Z188743 Wheaton sample vials, clear, volume 24 mL, screw-cap size, 20 ‑ 400, diam. × H 23 mm × 86 mm
Z256196 Wheaton sample vials with rubber lined caps, clear glass vial, volume 16 mL, thread 18-400
Z188735 Wheaton sample vials, clear, volume 16 mL, screw-cap size, 18 ‑ 400, diam. × H 21 mm × 71 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?