EMAIL THIS PAGE TO A FRIEND
Z252433 Aldrich
Wheaton vial rack
20 mL scintillation vials up to 28 mm diameter and 40 mL CombiChem/EPA vials, no. of holes, 50 (5 × 10)
Purchase
Properties
Related Categories Alcan Packaging, Alcan Packaging Vials & Caps, Analytical/Chromatography, Labware, Manufacturer Browser,
feature   no. of holes 50 (5 × 10)
packaging   pkg of 5 ea
L × W × H   17 cm × 32 cm × 3 cm
Description
General description
Chemically resistant polypropylene racks have corner posts to ensure stability when stacking empty. Autoclavable at 121°C for 20min. Vial grid is alphanumerically indexed for easy sample identification.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
V6880 Liquid scintillation vials, plastic, volume 20 mL, cap size 22 mm , polypropylene with metal foil liner, vial size 27 mm × 61 mm , with cap, pkg of 500 ea
V7005 Liquid scintillation vials, glass, volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with Poly-Seal cone liner. (caps packed separately), pkg of 500 ea
Z190543 Liquid scintillation vials (with screw cap attached), urethane foam-filled high-density polyethylene cap, liner (Poly-Seal cone), pkg of 500 ea
V8255 Liquid scintillation vials, glass, volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with metal foil liner. (caps packed separately), pkg of 500 ea
V6755 Liquid scintillation vials, plastic, volume 20 mL, cap size 24 mm , polyethylene without liner, vial size 27 mm × 61 mm , with cap, pkg of 1000 ea
Z253081 Liquid scintillation vials, volume 20 mL, clear glass vial, vials only, O.D. × H 28 mm × 57.5 mm, thread 22-400, pkg of 500 ea
Z190527 Liquid scintillation vials (with screw cap attached), polypropylene cap, foamed PE liner, pkg of 500 ea
V7130 Liquid scintillation vials, glass, volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with Poly-Seal liner., pkg of 500 ea
Z190519 Liquid scintillation vials (with screw cap attached), urethane foam-filled high-density polyethylene cap, metal foil liner, pkg of 500 ea
Z190535 Liquid scintillation vials (with screw cap attached), white polypropylene cap, metal foil liner, pkg of 500 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?