EMAIL THIS PAGE TO A FRIEND
Z252441 Aldrich
Wheaton vial rack
16-17mm culture tubes and 1.4-8 mL vials, no. of holes, 90 (6 × 15)
Purchase
Properties
Related Categories Alcan Packaging, Alcan Packaging Vials & Caps, Analytical/Chromatography, Labware, Manufacturer Browser,
feature   no. of holes 90 (6 × 15)
packaging   pkg of 5 ea
L × W × H   17 cm × 32 cm × 3 cm
Description
General description
Chemically resistant polypropylene racks have corner posts to ensure stability when stacking empty. Autoclavable at 121°C for 20min. Vial grid is alphanumerically indexed for easy sample identification.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
23202 Space Saver storage racks, for use with 17 mm diameter vial (90 vials per rack), well opening 17.1 mm, pkg of 5 ea
related product
Product #
Description
Add to Cart
Z256269 Wheaton sample vials with rubber lined caps, clear glass vial (white marking area), volume 8 mL, thread 15-425
Z188794 Wheaton sample vials, amber, volume 8 mL, screw-cap size, 15 ‑ 425, diam. × H 17 mm × 61 mm
Z256161 Wheaton sample vials with rubber lined caps, clear glass vial, volume 8 mL, thread 15-425
Z256293 Wheaton sample vials with rubber lined caps, amber glass vial, volume 8 mL, thread 15-425
Z188719 Wheaton sample vials, clear, volume 8 mL, screw-cap size, 15 ‑ 425, diam. × H 17 mm × 61 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?