EMAIL THIS PAGE TO A FRIEND
Z252484 Aldrich
Wheaton vial rack
12 mm vial, no. of holes, 50 (5 × 10)
Purchase
Properties
Related Categories Alcan Packaging, Alcan Packaging Vials & Caps, Analytical/Chromatography, Labware, Manufacturer Browser,
feature   no. of holes 50 (5 × 10)
packaging   pkg of 5 ea
L × W × H   19 cm × 10 cm × 2.2 cm
Description
General description
Chemically resistant polypropylene racks have corner posts to ensure stability when stacking empty. Autoclavable at 121°C for 20min. Vial grid is alphanumerically indexed for easy sample identification.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
23207 Space Saver storage racks, for use with 11-12 mm diameter vial (50 vials per rack), well opening 12.5 mm, pkg of 5 ea
related product
Product #
Description
Add to Cart
Z291633 Standard screw-thread autosampler vials, 12 × 32 mm, volume 2 mL, clear glass vial (standard opening), thread 8-425
Z162523 Vials, screw top, polypropylene, volume 0.3 mL, O.D. × H 12 mm × 32 mm, thread 8-425, pkg of 1000 ea
Z291641 Standard screw-thread autosampler vials, 12 × 32 mm, volume 2 mL, amber glass vial (standard opening), thread 8-425
Z291587 Autosampler crimp top vials 12 x 32mm standard opening, volume 2 mL, amber glass
Z188778 Wheaton sample vials, amber, volume 2 mL, screw-cap size, 8 ‑ 425, diam. × H 12 mm × 36 mm
Z256056 Wheaton sample vials with rubber lined caps, clear glass vial, volume 2 mL, thread 8-425
Z256277 Wheaton sample vials with rubber lined caps, volume 2 mL, amber glass vial, thread 8-425
Z162515 Wheaton PP autosampler vials, Snap Ring, volume 0.1 mL, natural polypropylene vial, 5 × 11 serum finish, thread 11 mm
Z188697 Wheaton sample vials, clear, volume 2 mL, screw-cap size, 8 ‑ 425, diam. × H 12 mm × 36 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?