• USA Home
  • Z252662 - Oxford Chemistry Primers
EMAIL THIS PAGE TO A FRIEND
Z252662 Aldrich
Oxford Chemistry Primers
No. 23: Inorganic Materials Chemistry
  •  ISBN-10 0198559981
  •  ISBN-13 978-019855998-6
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   M. Weller, Oxford University Press, 1995, 94 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
Although the chemistry of solid inorganic materials has become increasingly central to chemistry research, the subject has long been inadequately covered. This inexpensive and well-illustrated primer fills the gap with a comprehensive introduction to the subject. Topics include: transition metal oxides, non-stoichiometry, zeolites, layer compounds chemistry, high temperature superconductors, and fullerides. Syntheses of these compound types are presented as well. The book also details powder x-ray diffraction, the major experimental technique used to study and characterize the chemistry of solid inorganic materials. The basis for the method and associated, relevant crystallography is discussed. The experimental data gleaned from the technique is used to illustrate key topics throughout the primer. This tying together of an experimental method and the chemistry is a much needed approach, reflecting the true need of the chemist and the method by which chemistry is taught
Table of Contents
1. Basic Crystallography
2. Application and Interpretation of Powder x-ray Diffraction Data
3. The Synthesis of Inorganic Materials
4. Transition Metal Oxides
5. Electronic, Magnetic and Optical Properties of Inorganic Materials
6. Non-stoichiometry
7. Zeolites, Intercalation in Layer Materials and Solid Electrolytes
8. Some Recent Developments
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?