• USA Home
  • Z252689 - Tellurium In Organic Synthesis (Best Synthetic Methods)
EMAIL THIS PAGE TO A FRIEND
Z252689 Aldrich
Tellurium In Organic Synthesis (Best Synthetic Methods)
  •  ISBN-10 0125528108
  •  ISBN-13 978-012552810-8
Purchase
Properties
publication info   N. Petragnani, ed., Academic Press, 1994, 248 pp., hard cover
mfr. no.   Elsevier
Description
General description
The main purpose of this book is to help the practitioner prepare selected inorganic tellurium reagents and organotellurium compounds, and to apply these reagents in organic synthesis. The text discusses in detail the most significant reactions performed with tellurium reagents, including selective oxidations, selective reductions, the removal of protecting groups, and organotellurides and carbon radical scavengers, all under mild conditions.
Table of Contents
Preparation of the More Important Inorganic Tellurium Reagents.
The Preparation of the Principal Classes of Organic Tellurium Compounds.
Tellurium in Organic Synthesis.
Miscellanea.
Subject Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?