• USA Home
  • Z252786 - “Too Hot” safety labels
EMAIL THIS PAGE TO A FRIEND
Z252786 Aldrich
“Too Hot” safety labels
size 18 mm × 18 mm, pkg of 40 ea
Purchase
Properties
Related Categories General Labels, Labels and Markers, Labware, Pre-Printed Labels
packaging   pkg of 40 ea
size   18 mm × 18 mm
Description
General description
Self-adhesive, plastic-coated, waterproof labels are printed with special reversible liquid crystal inks. Label is black when cool, turns bright orange when "too hot" to touch, and reverts to black when cooled down. Use on glassware, water baths, hot plates, steam pipes, etc., to give instant visual warning and reduce burn accidents in the laboratory. Warning color change is at 47 °C, maximum use temperature is 100 °C.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?