EMAIL THIS PAGE TO A FRIEND
Z252867 Aldrich
Qwik Cell kit
Purchase
Description
General description
Obtain qualitative spectra on liquids and mulls with extended spectral range (to approx. 360cm-1). The kit consists of a Delrin® Qwik Cell body with screw top, six 15.8 × 0.5mm AgCl windows, assorted PTFE spacers (0.025, 0.05, and 0.1mm), and a cell holder.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?