• USA Home
  • Z253294 - Oakton® dual timer/clock
EMAIL THIS PAGE TO A FRIEND
Z253294 Aldrich
Oakton® dual timer/clock
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Oakton, Timers
mfr. no.   Oakton
W × H × D   5 1/2 in. × 3 3/4 in. × 5/8 in.
Description
General description
Large, tilting, dual LCD is readable from across the room. Two channels operate and display simultaneously to count up to, down from, or up to and down from 23h, 59min, 59s. Alarm sounds at end of countdown. Timer recalls previously set time. Built-in clock displays time in 12 or 24h format. Tabletop or wall-mountable. Includes one AAA battery.
Legal Information
Oakton is a registered trademark of Cole-Parmer Instrument Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z672076 Count up/down timer clock, complete with a pocket clip, foldaway stand and magnetic back; desk mountable
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?