• USA Home
  • Z253456 - Organic Synthesis, Volume 71
EMAIL THIS PAGE TO A FRIEND
Z253456 Aldrich
Organic Synthesis, Volume 71
  •  ISBN-10 0471305316
  •  ISBN-13 978-047130531-6
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   L. Overman, ed., John Wiley & Sons, Ltd, 1993, 320 pp., hard cover
mfr. no.   Wiley
Description
General description
Consists of 30 checked and edited experimental procedures that illustrate new synthetic techniques or describe the preparation of particularly useful chemicals. Features procedures exemplifying important methods for preparing enantiomerically pure substances by asymmetric catalysis; details a reliable zirconium-based process for the coupling of alkynes and enones; concludes with the preparation of four useful starting materials. A section on waste disposal information is included.
Table of Contents
PARTIAL TABLE OF CONTENTS:
Asymmetric Hydrogenation of 3-OXO Carboxylates Using Binapruthenium Complexes: (R)-(-)-Methyl 3-Hydroxybutanoate.
Direct Degradation of the Biopolymer Poly [(R)-3-Hydroxybutyric Acid to (R)-3-Hydroxybutanoic Acid and Its Methyl Ester.
3-(S)-[(tert-Butyldiphenylsilyl)-Oxy-2-Butanone.
Schwartz′s Reagent.
4-Methoxy-4′-Nitrobiphenyl.
Ubiquinone-1.
Benzoannelation of Ketones: 3,4-Cyclododeceno-1-Methylbenzene.
2-Methylene-1, 3-Dithiolane.
Methoxycarbonylmethylation of Aldehydes Via Siloxycyclopropanes: Methyl 3,3-Dimethyl-4-Oxobutanoate.
9-Bromo-9-Phenylfluorene.
Indexes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?