• USA Home
  • Z255831 - Latex rubber laboratory tubing
EMAIL THIS PAGE TO A FRIEND
Z255831 Aldrich
Latex rubber laboratory tubing
I.D. × O.D. 1/4 in. × 1/2 in.
Purchase
Properties
Related Categories Labware, Rubber Tubing, Tubing More...
description   standard
packaging   pkg of 100 feet
I.D. × O.D.   1/4 in. × 1/2 in.
wall size   1/8 in.
Description
General description
High-purity, seamless, amber tubing for general-purpose use with liquids and gases. Durometer: 35 ±5. Talc-free.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?